Assay Details
Target Gene Details
Entrez Gene ID: | 102238433 |
Gene Name: | predicted gene, 28042 |
Gene Aliases: |
Jmjd7-pla2g4b |
Location: |
Chr.2:120027483-120043033 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm28042 | NR_104353.1 | |||
AB195274.1 | ||||
AB195275.1 |
Target Gene Details
Entrez Gene ID: | 211429 |
Gene Name: | phospholipase A2, group IVB (cytosolic) |
Gene Aliases: |
A030011C02Rik, Gm1013, cPLA2-beta |
Location: |
Chr.2:120033433-120043033 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2g4b | NM_145378.4 | NP_663353.3 | ||
AK037218.1 | ||||
AK137721.1 | ||||
DQ888308.1 |
More Information
Set Membership: |
Intragenic Non-exonic |