Assay Details
Target Gene Details
Entrez Gene ID: | 241656 |
Gene Name: | p21 protein (Cdc42/Rac)-activated kinase 7 |
Gene Aliases: |
2900083L08Rik, 6430627N20, Pak5 |
Location: |
Chr.2:136081088-136387967 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pak7 | NM_172858.2 | 6 | 2031 | NP_766446.2 |
AK032593.1 | 6 | 2004 | BAC27939.1 | |
AK079080.1 | 7 | 2124 | BAC37528.1 | |
AK163530.1 | 6 | 1839 | BAE37385.1 | |
AY487425.1 | 4 | 1551 | AAR37415.1 |
More Information
Set Membership: |
Intragenic Exonic |