Assay Details
Target Gene Details
Entrez Gene ID: | 18286 |
Gene Name: | outer dense fiber of sperm tails 2 |
Gene Aliases: |
AI848335, MMTEST29 |
Location: |
Chr.2:29889720-29931746 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Odf2 | NM_001113213.1 | 5 | 691 | NP_001106684.1 |
NM_001113214.1 | 5 | 691 | NP_001106685.1 | |
NM_001177659.1 | 5 | 691 | NP_001171130.1 | |
NM_001177661.1 | 4 | 537 | NP_001171132.1 | |
NM_013615.3 | 5 | 664 | NP_038643.1 | |
AF000968.1 | 3 | 511 | AAB65157.1 | |
AF034105.1 | 5 | 627 | AAB87525.1 | |
AK143726.1 | 5 | 595 | BAE25517.1 | |
AK148523.1 | 5 | 604 | BAE28599.1 | |
AK165588.1 | 5 | 595 | ||
AK168415.1 | 5 | 593 | BAE40330.1 | |
BC057001.1 | 6 | 748 | AAH57001.1 | |
BQ840288.1 | 3 | 249 | ||
DQ091767.1 | 5 | 536 | AAZ39867.1 | |
DQ091770.1 | 5 | 595 | AAZ39870.1 |