Assay Details
Target Gene Details
Entrez Gene ID: | 107586 |
Gene Name: | ovo-like 2 (Drosophila) |
Gene Aliases: |
1700108N11Rik, 1810007D21Rik, M-OVO, M-OVO-A, M-OVO-B, MOVO, Ovo2, Zfp339, movo2 |
Location: |
Chr.2:144305175-144332080 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ovol2 | NM_026924.3 | NP_081200.2 | ||
NM_152947.2 | NP_694455.2 | |||
AB101294.1 | BAD29939.1 | |||
AB101295.1 | BAD29940.1 | |||
AB101296.1 | BAD29941.1 | |||
AK007360.1 | BAB24985.1 | |||
AK167697.1 | BAE39742.1 | |||
AY090537.1 | AAM11991.1 | |||
AY090538.1 | AAM11992.1 | |||
BC031771.1 | AAH31771.1 | |||
BC057210.1 | AAH57210.1 | |||
BC094445.1 | AAH94445.1 | |||
BY098454.1 |