Assay Details
Target Gene Details
Entrez Gene ID: | 64685 |
Gene Name: | N-myc (and STAT) interactor |
Gene Aliases: |
- |
Location: |
Chr.2:51948498-51973520 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nmi | NM_001141948.1 | NP_001135420.1 | ||
NM_001141949.1 | NP_001135421.1 | |||
NM_019401.2 | NP_062274.1 | |||
AF019249.1 | AAB70819.1 | |||
AK146577.1 | BAE27273.1 | |||
BC002019.1 | AAH02019.1 | |||
BY013052.1 | ||||
CJ093824.1 | ||||
CT010361.1 | CAJ18568.1 |
More Information
Set Membership: |
Intragenic Non-exonic |