Assay Details
Target Gene Details
Entrez Gene ID: | 50884 |
Gene Name: | NCK-associated protein 1 |
Gene Aliases: |
C79304, H19, Hem-2, Hem2, Nap1, mKIAA0587, mh19, p125Nap1 |
Location: |
Chr.2:80500512-80581259 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nckap1 | NM_001290745.1 | 10 | 1116 | NP_001277674.1 |
NM_016965.3 | 9 | 1098 | NP_058661.1 | |
AJ534525.1 | 9 | 881 | CAD58932.1 | |
AK122318.1 | 8 | 919 | BAC65600.1 | |
AK195089.1 | 4 | 318 | ||
BC127067.1 | 10 | 898 | ||
X61453.1 | CAA43693.1 |