Assay Details
Target Gene Details
Entrez Gene ID: | 50884 |
Gene Name: | NCK-associated protein 1 |
Gene Aliases: |
C79304, H19, Hem-2, Hem2, Nap1, mKIAA0587, mh19, p125Nap1 |
Location: |
Chr.2:80500512-80581259 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 26 - Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nckap1 | NM_001290745.1 | NP_001277674.1 | ||
NM_016965.3 | NP_058661.1 | |||
AJ534525.1 | CAD58932.1 | |||
AK122318.1 | BAC65600.1 | |||
AK143049.1 | BAE25258.1 | |||
BC003962.1 | AAH03962.1 | |||
BC127067.1 | ||||
X61453.1 | CAA43693.1 |