Assay Details
Target Gene Details
Entrez Gene ID: | 228019 |
Gene Name: | methyltransferase like 8 |
Gene Aliases: |
BC004636, Tip |
Location: |
Chr.2:70964558-71055643 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mettl8 | NM_001110512.1 | NP_001103982.1 | ||
NM_145524.3 | NP_663499.2 | |||
AK087927.1 | ||||
BC004636.1 | AAH04636.1 | |||
BC057960.1 | AAH57960.1 | |||
DQ093357.1 | AAZ04166.1 | |||
DQ093358.1 | AAZ04167.1 | |||
DQ230973.1 | ABB54392.1 | |||
DQ230974.1 | ABB54393.1 |
More Information
Set Membership: |
Intragenic Non-exonic |