Assay Details
Target Gene Details
Entrez Gene ID: | 16420 |
Gene Name: | integrin beta 6 |
Gene Aliases: |
2210409C20Rik, 4831415H04Rik |
Location: |
Chr.2:60598292-60722603 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itgb6 | NM_001159564.1 | NP_001153036.1 | ||
NM_021359.3 | NP_067334.1 | |||
AF115376.1 | AAD17212.1 | |||
AK008867.1 | 8 | 1341 | ||
AK036439.1 | BAC29430.1 | |||
AK085900.1 | ||||
BC049185.1 | AAH49185.1 |
More Information
Set Membership: |
Intragenic Non-exonic |