Assay Details
Target Gene Details
Entrez Gene ID: | 14571 |
Gene Name: | glycerol phosphate dehydrogenase 2, mitochondrial |
Gene Aliases: |
AA408484, AI448216, AU021455, AW494132, GPDH, Gdm1, Gpd-m, Gpdh-m, TISP38 |
Location: |
Chr.2:57237678-57370719 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpd2 | NM_001145820.1 | 7 | 1057 | NP_001139292.1 |
NM_010274.3 | 7 | 932 | NP_034404.3 | |
AK034353.1 | 7 | 841 | BAC28685.1 | |
AK144716.1 | 7 | 1058 | BAE26028.1 | |
AK151189.1 | 7 | 844 | ||
AK167152.1 | 7 | 870 | BAE39294.1 | |
BC021359.1 | 7 | 1026 | AAH21359.1 | |
D50430.1 | 7 | 834 | BAA08926.1 | |
U60987.1 | 7 | 868 | AAB50545.1 |
More Information
Set Membership: |
Intragenic Exonic |