Assay Details
Target Gene Details
Entrez Gene ID: | 107435 |
Gene Name: | histone aminotransferase 1 |
Gene Aliases: |
2410071B14Rik, AA536933, Hat-1, KAT1 |
Location: |
Chr.2:71389260-71441622 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hat1 | NM_026115.4 | 9 | 887 | NP_080391.2 |
AK010715.1 | 9 | 897 | ||
AK014330.1 | 9 | 898 | ||
AK018270.1 | 9 | 878 | ||
AK041700.1 | 9 | 891 | BAC31038.1 | |
AK169315.1 | 9 | 890 | BAE41070.1 | |
BC055460.1 | 8 | 854 | AAH55460.1 |
More Information
Set Membership: |
Intragenic Exonic |