Assay Details
Target Gene Details
Entrez Gene ID: | 14683 |
Gene Name: | GNAS (guanine nucleotide binding protein, alpha stimulating) complex locus |
Gene Aliases: |
5530400H20Rik, A930027G11Rik, C130027O20Rik, GPSA, GSP, Galphas, Gnas1, Gnasxl, Gsa, Nesp, Nespl, Oed-Sml, Oedsml, P1, P2, P3, PHP1A, PHP1B, POH |
Location: |
Chr.2:174283444-174346744 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gnas | NM_019690.2 | NP_062664.2 | ||
NM_022000.2 | NP_068840.2 | |||
AF107847.1 | AAD11805.1 | |||
AF107848.1 | AAD11806.1 | |||
AF175305.1 | AAD55061.1 | |||
AJ010163.1 | CAA09026.1 | |||
AK039035.1 | BAC30214.1 | |||
AK161197.1 |
Target Gene Details
Entrez Gene ID: | 56802 |
Gene Name: | neuroendocrine secretory protein antisense |
Gene Aliases: |
Gnas-as, Nespos |
Location: |
Chr.2:174281237-174295436 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nespas | NR_002846.2 | |||
AF173359.1 | ||||
AF319945.1 | ||||
AK153813.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |