Assay Details
Target Gene Details
Entrez Gene ID: | 66691 |
Gene Name: | GTPase activating protein and VPS9 domains 1 |
Gene Aliases: |
2010005B09Rik, 4432404J10Rik, AW108497, Gapex-5, RAP6, RME-6, mKIAA1521 |
Location: |
Chr.2:34676179-34755263 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gapvd1 | NM_025709.2 | 12 | 2629 | NP_079985.2 |
AK129381.1 | 11 | 2224 | BAC98191.1 | |
AK134776.1 | 10 | 1619 | BAE22277.1 | |
AK150026.1 | 11 | 2238 | BAE29251.1 | |
BC031478.1 | 9 | 1674 | ||
BC048847.1 | 11 | 2195 | AAH48847.1 | |
BC060123.1 | 12 | 2629 | AAH60123.1 | |
EF155419.1 | 10 | 2047 |
More Information
Set Membership: |
Intragenic Exonic |