Assay Details
Target Gene Details
Entrez Gene ID: | 66691 |
Gene Name: | GTPase activating protein and VPS9 domains 1 |
Gene Aliases: |
2010005B09Rik, 4432404J10Rik, AW108497, Gapex-5, RAP6, RME-6, mKIAA1521 |
Location: |
Chr.2:34676179-34755263 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gapvd1 | NM_025709.2 | 10 | 2316 | NP_079985.2 |
AK129381.1 | 9 | 1911 | BAC98191.1 | |
AK134776.1 | 8 | 1306 | BAE22277.1 | |
AK150026.1 | 9 | 1925 | BAE29251.1 | |
BC031478.1 | 7 | 1361 | ||
BC048847.1 | 9 | 1882 | AAH48847.1 | |
BC060123.1 | 10 | 2316 | AAH60123.1 | |
EF155419.1 | 8 | 1734 |
More Information
Set Membership: |
Intragenic Exonic |