Assay Details
Target Gene Details
Entrez Gene ID: | 99138 |
Gene Name: | START domain containing 7 |
Gene Aliases: |
AI852671, AL022671, AW544915 |
Location: |
Chr.2:127270203-127298935 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Stard7 | NM_139308.2 | 1 | 243 | NP_647469.2 |
AK143699.1 | 1 | 244 | BAE25505.1 | |
BC017524.1 | 1 | 233 | ||
BC080747.1 | 1 | 214 | AAH80747.1 |