Assay Details
Target Gene Details
Entrez Gene ID: | 14269 |
Gene Name: | formin binding protein 1 |
Gene Aliases: |
1110057E06Rik, 2210010H06Rik, FBP1, Fbp17 |
Location: |
Chr.2:31026206-31142027 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fnbp1 | NM_001038700.2 | NP_001033789.1 | ||
NM_001177648.1 | NP_001171119.1 | |||
NM_001177649.1 | NP_001171120.1 | |||
NM_001177650.1 | NP_001171121.1 | |||
NM_019406.3 | NP_062279.1 | |||
AK122308.1 | BAC65590.1 | |||
AK143512.1 | BAE25408.1 | |||
AK157135.1 | BAE33974.1 | |||
AK170497.1 | BAE41836.1 | |||
AK172100.1 | BAE42825.1 | |||
BC003867.1 | AAH03867.1 |
More Information
Set Membership: |
Intragenic Non-exonic |