Assay Details
Target Gene Details
Entrez Gene ID: | 77683 |
Gene Name: | euchromatic histone methyltransferase 1 |
Gene Aliases: |
9230102N17Rik, D330003E03, Eu-HMTase1, GLP, GLP1, KMT1D, mKIAA1876 |
Location: |
Chr.2:24790769-24919643 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ehmt1 | NM_001012518.3 | 15 | 2365 | NP_001012536.2 |
NM_001109686.2 | 14 | 2206 | NP_001103156.1 | |
NM_001109687.2 | 14 | 2221 | NP_001103157.1 | |
NM_172545.4 | 15 | 2344 | NP_766133.2 | |
AB205007.1 | 15 | 2371 | BAD90007.1 | |
AK049454.1 | 12 | 1615 | BAC33756.1 | |
AK052174.1 | 15 | 2328 | BAC34869.1 | |
AK054555.1 | 15 | 2313 | ||
AK082062.1 | 15 | 2355 | BAC38402.1 | |
BC056938.1 | 12 | 1922 | AAH56938.1 | |
BC089302.1 | 12 | 2074 | AAH89302.1 |