Assay Details
Target Gene Details
Entrez Gene ID: | 66861 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 10 |
Gene Aliases: |
1200006L06Rik, D2Ertd706e, ERdj5, JPDI |
Location: |
Chr.2:80315466-80354055 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dnajc10 | NM_024181.2 | 6 | 1047 | NP_077143.2 |
AF255459.1 | 6 | 724 | AAN73273.1 | |
AF314002.1 | 5 | 560 | AAQ14555.1 | |
AK004617.1 | 6 | 802 | BAB23413.1 | |
AK028184.1 | 6 | 787 | ||
AK128945.1 | 6 | 802 | ||
AK165652.1 | 6 | 805 | ||
AK166502.1 | 6 | 757 | ||
AK169020.1 | 6 | 814 | ||
AK179556.1 | 4 | 296 | ||
BC033461.1 | 6 | 786 | AAH33461.1 |
More Information
Set Membership: |
Intragenic Exonic |