Assay Details
Target Gene Details
Entrez Gene ID: | 107986 |
Gene Name: | damage specific DNA binding protein 2 |
Gene Aliases: |
2610043A19Rik |
Location: |
Chr.2:91211579-91237132 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddb2 | NM_028119.5 | NP_082395.2 | ||
AK011756.1 | ||||
AK158770.1 | BAE34654.1 | |||
AY027937.1 | AAK16810.1 | |||
BC099410.1 | AAH99410.1 | |||
BU515611.1 |
More Information
Set Membership: |
Intragenic Non-exonic |