Assay Details
Target Gene Details
Entrez Gene ID: | 66953 |
Gene Name: | cell division cycle associated 7 |
Gene Aliases: |
2310021G01Rik, AU044109, JPO1 |
Location: |
Chr.2:72476189-72486892 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdca7 | NM_025866.3 | NP_080142.1 | ||
AK011289.1 | BAB27519.1 | |||
AK028671.1 | BAC26058.1 | |||
AK075882.1 | BAC36027.1 | |||
AK153943.1 | BAE32269.1 | |||
AK167734.1 | BAE39775.1 | |||
BC066169.1 | AAH66169.1 |