Assay Details
Target Gene Details
Entrez Gene ID: | 70266 |
Gene Name: | cysteine conjugate-beta lyase 1 |
Gene Aliases: |
2010009K05Rik, AI182306, Gtk, Kat1, KatI |
Location: |
Chr.2:30185124-30206295 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccbl1 | NM_172404.2 | NP_765992.2 | ||
AK008165.1 | ||||
AK031511.1 | ||||
AK042391.1 | BAC31248.1 | |||
AK088404.1 | BAC40333.1 | |||
AK163165.1 | ||||
AK207151.1 | ||||
AK218327.1 | ||||
BC016206.1 | AAH16206.1 | |||
BC025124.1 | ||||
BC052047.1 | AAH52047.1 |
More Information
Set Membership: |
Intragenic Non-exonic |