Assay Details
Target Gene Details
Entrez Gene ID: | 252966 |
Gene Name: | CDK5 and Abl enzyme substrate 2 |
Gene Aliases: |
ik3-2 |
Location: |
Chr.2:180258539-180273490 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cables2 | NM_145851.2 | NP_665850.2 | ||
AK050121.1 |