Assay Details
Target Gene Details
Entrez Gene ID: | 67512 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 2 (lysophosphatidic acid acyltransferase, beta) |
Gene Aliases: |
2510002J07Rik, AV000834, BSCL, BSCL1, LPAAB, LPAAT-beta |
Location: |
Chr.2:26593057-26604417 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Agpat2 | NM_026212.2 | NP_080488.1 | ||
AK010891.1 | ||||
AK090271.1 | ||||
AY072769.1 | AAL62337.1 | |||
BC125530.1 | ||||
BC137840.1 |
More Information
Set Membership: |
Intragenic Non-exonic |