Assay Details
Target Gene Details
Entrez Gene ID: | 11432 |
Gene Name: | acid phosphatase 2, lysosomal |
Gene Aliases: |
Acp-2, LAP |
Location: |
Chr.2:91202812-91214098 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acp2 | NM_007387.2 | NP_031413.1 | ||
AK089401.1 | ||||
AK128963.1 | ||||
AK133756.1 | BAE21825.1 | |||
AK154269.1 | BAE32478.1 | |||
AK170658.1 | ||||
BC023343.1 | AAH23343.1 | |||
X57199.1 | CAA40485.1 |
More Information
Set Membership: |
Intragenic Non-exonic |