Assay Details
Target Gene Details
Entrez Gene ID: | 215456 |
Gene Name: | glycerol-3-phosphate acyltransferase 2, mitochondrial |
Gene Aliases: |
A530057A03Rik, Gm116, xGPAT1 |
Location: |
Chr.2:127410628-127436093 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpat2 | NM_001081089.2 | 13 | 1390 | NP_001074558.2 |
AB257502.1 | 12 | 1308 | ||
AK131154.1 | 13 | 1389 | BAD21404.1 | |
BC113165.1 | 13 | 1384 | ||
BC113776.1 | 13 | 1381 |
More Information
Set Membership: |
Intragenic Exonic |