Assay Details
Target Gene Details
Entrez Gene ID: | 11350 |
Gene Name: | c-abl oncogene 1, non-receptor tyrosine kinase |
Gene Aliases: |
AI325092, Abl, E430008G22Rik, c-Abl |
Location: |
Chr.2:31688252-31807093 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abl1 | NM_001112703.2 | NP_001106174.1 | ||
NM_001283045.1 | NP_001269974.1 | |||
NM_001283046.1 | NP_001269975.1 | |||
NM_001283047.1 | NP_001269976.1 | |||
NM_009594.4 | NP_033724.2 | |||
AK088252.1 | BAE43394.1 | |||
AK090095.1 | BAC41088.1 | |||
AK166028.1 | ||||
AK166210.1 | BAE38629.1 | |||
BC059260.1 | AAH59260.1 | |||
BC103770.1 | AAI03771.1 | |||
CF726668.1 | ||||
J02995.1 | AAA88241.1 | |||
L10656.1 |