Assay Details
Target Gene Details
Entrez Gene ID: | 75763 |
Gene Name: | DDB1 and CUL4 associated factor 17 |
Gene Aliases: |
2810055O12Rik, 4833418A01Rik, A030004A10Rik, A930009G19Rik, AI448937 |
Location: |
Chr.2:71055742-71114514 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dcaf17 | NM_001165980.1 | 10 | 1293 | NP_001159452.1 |
NM_001165981.1 | 10 | 1293 | NP_001159453.1 | |
NM_198005.2 | 10 | 1293 | NP_932122.2 | |
AK014717.1 | 10 | 1257 | ||
AK079462.1 | 10 | 1087 | BAC37654.1 | |
AK081839.1 | 10 | 1292 | ||
AK160679.1 | 10 | 1260 | BAE35954.1 | |
BC031546.1 | 10 | 1185 | AAH31546.1 |