Assay Details
Target Gene Details
Entrez Gene ID: | 12503 |
Gene Name: | CD247 antigen |
Gene Aliases: |
4930549J05Rik, A430104F18Rik, AW552088, Cd3, Cd3-eta, Cd3-zeta, Cd3h, Cd3z, Cd3zeta, T3z, Tcrk, Tcrz |
Location: |
Chr.1:165780357-165871153 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd247 | NM_001113392.2 | NP_001106863.1 | ||
NM_001113393.2 | NP_001106864.2 | |||
NM_031162.4 | 8 | 656 | NP_112439.1 | |
NR_103716.1 | 8 | 656 | ||
AK138774.1 | BAE23774.1 | |||
L03353.1 | AAA37401.1 | |||
M33158.1 | 8 | 646 | AAA37398.1 | |
S51932.1 | AAB24559.1 | |||
U56258.1 | 8 | 572 | AAD11545.1 | |
X84237.1 | CAA59015.1 |
Target Gene Details
Entrez Gene ID: | 18986 |
Gene Name: | POU domain, class 2, transcription factor 1 |
Gene Aliases: |
2810482H01Rik, NF-A1, Oct-1, Oct1, Otf-1, Otf1 |
Location: |
Chr.1:165865150-166002672 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pou2f1 | NM_011137.3 | 17 | 11399 | NP_035267.2 |
NM_198932.3 | 16 | 11327 | NP_945150.2 | |
NM_198933.3 | 16 | 11399 | NP_945151.2 | |
NM_198934.3 | 17 | 11471 | NP_945152.2 | |
AK051825.1 | 1 | 248 |
More Information
Set Membership: |
Intragenic Exonic |