Assay Details
Target Gene Details
Entrez Gene ID: | 107508 |
Gene Name: | glutamyl-prolyl-tRNA synthetase |
Gene Aliases: |
2410081F06Rik, 3010002K18Rik, C79379, Qprs |
Location: |
Chr.1:185363028-185428355 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eprs | NM_029735.1 | 19 | 2803 | NP_084011.1 |
AK028687.1 | 12 | 1824 | ||
AK141072.1 | 19 | 2858 | ||
AK148463.1 | 19 | 2764 | BAE28568.1 | |
AK160986.1 | 19 | 2823 | ||
BC040802.1 | 19 | 2801 | AAH40802.1 | |
BC094679.1 | 18 | 2597 | AAH94679.1 | |
BC141049.1 | 19 | 2722 | ||
X54327.1 | 10 | 1487 |
More Information
Set Membership: |
Intragenic Exonic |