Assay Details
Target Gene Details
Entrez Gene ID: | 66637 |
Gene Name: | tRNA splicing endonuclease 15 homolog (S. cerevisiae) |
Gene Aliases: |
5730449L18Rik, AL023077, Sen15 |
Location: |
Chr.1:152370735-152386682 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tsen15 | NM_025677.3 | NP_079953.2 | ||
AK004261.1 | ||||
AK011608.1 | ||||
AK017650.1 | BAB30855.1 | |||
AK169429.1 | 2 | 3049 | ||
BC024342.1 | AAH24342.1 | |||
BC145701.1 | ||||
BY065176.1 |