Assay Details
Target Gene Details
Entrez Gene ID: | 171382 |
Gene Name: | transient receptor potential cation channel, subfamily M, member 8 |
Gene Aliases: |
CMR1, LTRPC6, LTrpC-6, TRPP8, Trp-p8 |
Location: |
Chr.1:88306712-88388853 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trpm8 | NM_134252.3 | 16 | 2137 | NP_599013.1 |
AY095352.1 | 14 | 1971 | AAM23261.1 | |
BC117934.1 | 16 | 2137 |
More Information
Set Membership: |
Intragenic Exonic |