Assay Details
Target Gene Details
Entrez Gene ID: | 226438 |
Gene Name: | immunoglobulin-like and fibronectin type III domain containing 1 |
Gene Aliases: |
9830123M21Rik |
Location: |
Chr.1:135953205-136040842 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igfn1 |
Target Gene Details
Entrez Gene ID: | 66241 |
Gene Name: | transmembrane protein 9 |
Gene Aliases: |
1500015G18Rik, AW545782 |
Location: |
Chr.1:136008219-136035030 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem9 | NM_001160145.1 | 1 | 135 | NP_001153617.1 |
BQ918002.1 | 1 | 222 |