Assay Details
Target Gene Details
Entrez Gene ID: | 109232 |
Gene Name: | saccharopine dehydrogenase (putative) |
Gene Aliases: |
AW214504, C330023F11Rik |
Location: |
Chr.1:179668231-179687184 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sccpdh | NM_178653.3 | 4 | 567 | NP_848768.1 |
AK042919.1 | 4 | 455 | BAC31405.1 | |
AK046033.1 | 4 | 568 | BAC32578.1 | |
AK046468.1 | 4 | 554 | ||
AK050554.1 | 4 | 516 | ||
AK077695.1 | 4 | 517 | BAC36962.1 | |
AK082803.1 | 4 | 452 | BAC38628.1 | |
AK086448.1 | 4 | 590 | ||
AK086632.1 | 4 | 565 | ||
AK145399.1 | 4 | 551 | BAE26412.1 | |
AK166018.1 | 4 | 479 | BAE38523.1 | |
AK182882.1 | 4 | 309 | ||
AK215316.1 | 4 | 309 | ||
BC025803.1 | 4 | 564 | AAH25803.1 |