Assay Details
Target Gene Details
Entrez Gene ID: | 51801 |
Gene Name: | receptor (calcitonin) activity modifying protein 1 |
Gene Aliases: |
9130218E19Rik |
Location: |
Chr.1:91179822-91225196 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ramp1 | NM_001168392.1 | NP_001161864.1 | ||
NM_016894.3 | NP_058590.1 | |||
NM_178401.3 | NP_848488.2 | |||
AB019046.1 | BAA76617.1 | |||
AF146522.1 | AAD35018.1 | |||
AF209904.1 | AAF21036.1 | |||
AJ250489.1 | CAB59511.1 | |||
AK144331.1 | BAE25837.1 | |||
AK157896.1 | BAE34250.1 | |||
AK165238.1 | BAE38095.1 | |||
BC012644.1 | AAH12644.1 | |||
BY707433.1 | ||||
BY733497.1 | ||||
CB522711.1 |
More Information
Set Membership: |
Intragenic Non-exonic |