Assay Details
Target Gene Details
Entrez Gene ID: | 29809 |
Gene Name: | RAB GTPase activating protein 1-like |
Gene Aliases: |
5830411O09Rik, 8430421H08Rik, 9630005B12Rik, AW049894, HHL, Hh1, mKIAA0471 |
Location: |
Chr.1:160219174-160793476 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rabgap1l | NM_013862.5 | 10 | 1450 | NP_038890.3 |
AK034019.1 | 11 | 1367 | BAC28549.1 | |
AK035796.1 | 10 | 1451 | BAC29189.1 | |
BC145811.1 | 10 | 1295 | ||
BC145813.1 | 10 | 1295 |
More Information
Set Membership: |
Intragenic Exonic |