Assay Details
Target Gene Details
Entrez Gene ID: | 29809 |
Gene Name: | RAB GTPase activating protein 1-like |
Gene Aliases: |
5830411O09Rik, 8430421H08Rik, 9630005B12Rik, AW049894, HHL, Hh1, mKIAA0471 |
Location: |
Chr.1:160219174-160793476 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rabgap1l | NM_001038621.2 | 8 | 1043 | NP_001033710.1 |
AK018430.1 | 4 | 311 | BAB31207.1 | |
AK122292.1 | 6 | 538 | BAC65574.1 | |
AK147634.1 | 8 | 1044 | BAE28037.1 |
More Information
Set Membership: |
Intragenic Exonic |