Assay Details
Target Gene Details
Entrez Gene ID: | 18187 |
Gene Name: | neuropilin 2 |
Gene Aliases: |
1110048P06Rik, Np-2, Np2, Npn-2, Npn2 |
Location: |
Chr.1:62703317-62818692 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nrp2 | NM_001077403.1 | NP_001070871.1 | ||
NM_001077404.1 | NP_001070872.1 | |||
NM_001077405.1 | NP_001070873.1 | |||
NM_001077406.1 | NP_001070874.1 | |||
NM_001077407.1 | NP_001070875.1 | |||
NM_010939.2 | NP_035069.2 | |||
AF483506.1 | AAL90780.1 | |||
AF483507.1 | AAL90781.1 | |||
AK051873.1 | BAC34795.1 | |||
AK149643.1 | ||||
AK153553.1 | BAE32090.1 |
More Information
Set Membership: |
Intragenic Non-exonic |