Assay Details
Target Gene Details
Entrez Gene ID: | 12355 |
Gene Name: | nuclear receptor subfamily 1, group I, member 3 |
Gene Aliases: |
AA209988, AI551208, CAR, CAR-beta, Care2, ESTM32, MB67 |
Location: |
Chr.1:171213935-171218845 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nr1i3 | NM_001243062.1 | 2 | 274 | NP_001229991.1 |
NM_001243063.1 | 2 | 274 | NP_001229992.1 | |
NM_009803.5 | 2 | 274 | NP_033933.2 | |
AF009327.1 | 2 | 211 | AAC53349.1 | |
AF009328.1 | 2 | 308 | AAC53350.1 | |
AK133515.1 | 2 | 275 | BAE21697.1 | |
AK149381.1 | 2 | 226 | ||
BC037645.1 | 2 | 280 | AAH37645.1 | |
BC089541.1 | 2 | 223 | AAH89541.1 |