Assay Details
Target Gene Details
Entrez Gene ID: | 69527 |
Gene Name: | mitochondrial ribosomal protein S9 |
Gene Aliases: |
2310002A08Rik, AW322516, Mrp-S9, S9mt, mg637 |
Location: |
Chr.1:42851233-42905683 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mrps9 | NM_023514.4 | 1 | 51 | NP_076003.3 |
AK009082.1 | BAB26060.2 | |||
AK031497.1 | ||||
AK049465.1 | ||||
AK144656.1 | BAE25993.1 | |||
AK150985.1 | ||||
BC116437.2 | ||||
BY145802.1 | 1 | 51 |
More Information
Set Membership: |
Intragenic Exonic |