Assay Details
Target Gene Details
Entrez Gene ID: | 227157 |
Gene Name: | membrane protein, palmitoylated 4 (MAGUK p55 subfamily member 4) |
Gene Aliases: |
Dlg6 |
Location: |
Chr.1:59120935-59163389 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mpp4 | NM_001164682.1 | 11 | 1542 | NP_001158154.1 |
NM_145143.3 | 10 | 905 | NP_660125.2 | |
AB059357.1 | 9 | 796 | BAB69494.1 | |
AB059358.1 | 9 | 796 | BAB69495.1 | |
AJ748820.1 | 9 | 786 | CAG38657.1 | |
AK044682.1 | 11 | 1541 | BAC32031.1 | |
AK138845.1 | 7 | 2393 | ||
BC061694.1 | 10 | 822 | ||
BC116723.1 | 5 | 425 |
More Information
Set Membership: |
Intragenic Exonic |