Assay Details
Target Gene Details
Entrez Gene ID: | 65102 |
Gene Name: | Ngg1 interacting factor 3-like 1 (S. pombe) |
Gene Aliases: |
1110030G24Rik |
Location: |
Chr.1:58445156-58462276 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nif3l1 | NM_022988.2 | NP_075364.2 | ||
AF284439.1 | AAG45961.1 | |||
AK003978.1 | BAB23102.1 | |||
AK011670.1 | BAB27769.1 | |||
AK131631.1 | BAE20729.1 | |||
AK154815.1 | BAE32848.1 | |||
AK158616.1 | BAE34583.1 | |||
BC065163.1 | AAH65163.1 | |||
BC089475.1 | AAH89475.1 |