Assay Details
Target Gene Details
Entrez Gene ID: | 14768 |
Gene Name: | LanC (bacterial lantibiotic synthetase component C)-like 1 |
Gene Aliases: |
AW124738, Gpr69, Gpr69a, p40 |
Location: |
Chr.1:67000517-67038872 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lancl1 | NM_001190984.1 | 8 | 1002 | NP_001177913.1 |
NM_001190985.1 | 7 | 1220 | NP_001177914.1 | |
NM_021295.3 | 8 | 1036 | NP_067270.1 | |
AJ294535.1 | 8 | 1029 | CAC69241.1 | |
AK146120.1 | 7 | 1211 | ||
AK182820.1 | ||||
BC038649.1 | AAH38649.1 | |||
BC058560.1 | 8 | 1002 | AAH58560.1 | |
Y11550.1 | 8 | 1001 | CAA72314.1 | |
Y16518.1 | 7 | 1180 | CAA76269.1 |