Assay Details
Target Gene Details
Entrez Gene ID: | 26921 |
Gene Name: | mitogen-activated protein kinase kinase kinase kinase 4 |
Gene Aliases: |
9430080K19Rik, AU043147, AU045934, AW046177, HGK, Nik |
Location: |
Chr.1:39900912-40026310 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 25 - Intron 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Map4k4 | NM_001252200.1 | NP_001239129.1 | ||
NM_001252201.1 | NP_001239130.1 | |||
NM_001252202.1 | NP_001239131.1 | |||
NM_008696.2 | NP_032722.2 | |||
AK129191.1 | BAC98001.1 | |||
AK179068.1 | ||||
BC141109.1 | ||||
BC145390.1 | ||||
BC145391.1 | ||||
CX231827.1 | ||||
U88984.1 | AAC53165.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)