Assay Details
Target Gene Details
Entrez Gene ID: | 72017 |
Gene Name: | cytochrome b5 reductase 1 |
Gene Aliases: |
1500005G05Rik, B5R.1, C80155, Nqo3a2 |
Location: |
Chr.1:134405561-134411740 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyb5r1 | NM_028057.2 | 2 | 139 | NP_082333.1 |
AK005159.1 | 2 | 140 | BAB23850.1 | |
AK049717.1 | 1 | 361 | ||
AK159663.1 | 2 | 109 | BAE35270.1 | |
AK167178.1 | 2 | 133 | BAE39313.1 | |
BC016266.1 | 2 | 348 | AAH16266.1 | |
BC024618.1 | 2 | 117 | AAH24618.1 | |
JX420289.1 | 2 | 574 | ||
JX420290.1 | 2 | 316 |
More Information
Set Membership: |
Intragenic Exonic |