Assay Details
Target Gene Details
Entrez Gene ID: | 12958 |
Gene Name: | crystallin, beta A2 |
Gene Aliases: |
E130107M19Rik |
Location: |
Chr.1:74889934-74898843 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cryba2 | NM_021541.3 | 5 | 622 | NP_067516.1 |
AJ272227.1 | 5 | 614 | CAB75585.1 | |
AK021374.1 | 5 | 596 | BAB32389.1 | |
AY160973.1 | 5 | 587 | AAN78172.1 | |
BC052746.1 | 5 | 585 | AAH52746.1 | |
BC056989.1 | 5 | 576 | AAH56989.1 | |
CF740624.1 | 5 | 621 |