Assay Details
Target Gene Details
Entrez Gene ID: | 12958 |
Gene Name: | crystallin, beta A2 |
Gene Aliases: |
E130107M19Rik |
Location: |
Chr.1:74889934-74898843 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cryba2 | NM_021541.3 | NP_067516.1 | ||
AJ272227.1 | CAB75585.1 | |||
AK021374.1 | BAB32389.1 | |||
AY160973.1 | AAN78172.1 | |||
BC052746.1 | AAH52746.1 | |||
BC056989.1 | AAH56989.1 | |||
CF740624.1 |