Assay Details
Target Gene Details
Entrez Gene ID: | 21367 |
Gene Name: | contactin 2 |
Gene Aliases: |
D130012K04Rik, TAG-1, Tax |
Location: |
Chr.1:132509425-132543251 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cntn2 | NM_177129.5 | 3 | 456 | NP_796103.2 |
AK147573.1 | 3 | 382 | ||
AK147602.1 | 3 | 457 | BAE28018.1 | |
AK147658.1 | 3 | 382 | ||
AK147673.1 | 3 | 407 | BAE28065.1 | |
BC066106.1 | 3 | 376 | AAH66106.1 | |
CJ098637.1 | 3 | 457 | ||
CJ108593.1 | 3 | 457 |
More Information
Set Membership: |
Intragenic Exonic |