Assay Details
Target Gene Details
Entrez Gene ID: | 21367 |
Gene Name: | contactin 2 |
Gene Aliases: |
D130012K04Rik, TAG-1, Tax |
Location: |
Chr.1:132509425-132543251 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cntn2 | NM_177129.5 | NP_796103.2 | ||
AK147573.1 | ||||
AK147602.1 | BAE28018.1 | |||
AK147658.1 | ||||
AK147673.1 | BAE28065.1 | |||
BC053033.1 | AAH53033.1 | |||
BC066106.1 | AAH66106.1 |
More Information
Set Membership: |
Intragenic Non-exonic |