Assay Details
Target Gene Details
Entrez Gene ID: | 381319 |
Gene Name: | basic leucine zipper transcription factor, ATF-like 3 |
Gene Aliases: |
9130211I03Rik, Snft |
Location: |
Chr.1:191098414-191108943 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Batf3 | NM_030060.2 | NP_084336.1 | ||
AK020278.1 | BAB32053.1 | |||
BC117083.1 | ||||
BC117085.1 |
More Information
Set Membership: |
Intragenic Non-exonic |