Assay Details
Target Gene Details
Entrez Gene ID: | 66799 |
Gene Name: | ubiquitin-conjugating enzyme E2W (putative) |
Gene Aliases: |
6130401J04Rik |
Location: |
Chr.1:16540788-16619420 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ube2w | NM_001271016.1 | NP_001257945.1 | ||
NM_001271017.1 | NP_001257946.1 | |||
NM_025773.3 | NP_080049.2 | |||
NR_073122.1 | ||||
AK015348.1 | BAB29807.1 | |||
AK044463.1 | BAC31935.1 | |||
AK046086.1 | BAC32599.1 | |||
AK050679.1 | BAC34377.1 | |||
AK050923.1 | BAE43345.1 | |||
AK077997.1 | BAC37094.1 | |||
AK082233.1 | BAC38443.1 | |||
BC020124.1 | AAH20124.1 | |||
BY728312.1 | ||||
CN663576.1 | ||||
DV044691.1 |
More Information
Set Membership: |
Intragenic Non-exonic |